1-Methylimidazole (NMI) 98%

Supplier: Ambeed
Danger

Synonyms: N-Methylimidazole, 1-Methyl-1H-imidazole, N-Methyl-1H-imidazole, NMI

A918471-1000G A918471-100G A918471-25G A918471-500G
77118-866EA 46.68 USD
77118-866 77118-868 77118-870 77118-872
1-Methylimidazole (NMI) 98%
1-Methylimidazole

Caution: for Research Use Only

Formula: C₄H₆N₂
MW: 82.11 g/mol
Boiling Pt: 198
Melting Pt: -2
Flash Pt: 92
MDL Number: MFCD00005292
CAS Number: 616-47-7
UN: 2922
ADR: 8,6.1 Ⅱ

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR