3-Ethynyltoluene 97%
Supplier: Ambeed
|
Danger
|
A888384-10G
A888384-1G
A888384-25G
A888384-5G
77118-736EA
60.77
USD
77118-736
77118-738
77118-740
77118-742
3-Ethynyltoluene 97%
3-Ethynyltoluene
Caution: for Research Use Only
Formula:
C₉H₈ |
MDL Number:
MFCD01318174 CAS Number: 766-82-5 UN: 1993 ADR: 3,Ⅲ |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...