Verapamil hydrochloride 99%

Supplier: Ambeed
Danger

A724474-1G A724474-1MG A724474-5G A724474-25G A724474-100G
77118-060EA 28.82 USD
77118-060 77118-062 77118-064 77552-242 77686-722
Verapamil hydrochloride 99%
Verapamil hydrochloride

Caution: for Research Use Only

Formula: C₂₇H₃₈N₂O₄
MDL Number: MFCD00055208
CAS Number: 152-11-4
UN: 2811
ADR: 6.1,Ⅲ

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR