2'-Iodo-1,1':3',1''-terphenyl 95%

Supplier: Ambeed
Warning

Synonyms: 2'-Iodo-1,1':3',1''-terphenyl, 1-Iodo-2,6-diphenylbenzene, 2'-Iodo-m-terphenyl

A713061-100MG A713061-1G A713061-250MG A713061-5G
77117-996EA 25.75 USD
77117-996 77117-998 77118-000 77118-002
2'-Iodo-1,1':3',1''-terphenyl 95%
2'-Iodo-1,1':3',1''-terphenyl

Caution: for Research Use Only

Formula: C₁₈H₁₃I
MDL Number: MFCD00185026
CAS Number: 82777-09-1

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR