α-Bromo-2-(difluoromethoxy)toluene 98%

Supplier: Ambeed
Danger

A694064-1G A694064-25G A694064-5G
77117-894EA 22.66 USD
77117-894 77117-896 77117-898
α-Bromo-2-(difluoromethoxy)toluene 98%
α-Bromo-2-(difluoromethoxy)toluene

Caution: for Research Use Only

Formula: C₈H₇BrF₂O
MDL Number: MFCD00190103
CAS Number: 85684-64-6
UN: 3265
ADR: 8,Ⅱ

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR