(3-Bromo-2-methylphenyl)boronic acid 98%

Supplier: Ambeed
Warning

Synonyms: 3-Bromo-2-methylphenylboronic acid

A582580-100MG A582580-1G
77117-178EA 20.6 USD
77117-178 77117-180
(3-Bromo-2-methylphenyl)boronic acid 98%
(3-Bromo-2-methylphenyl)boronic acid

Caution: for Research Use Only

Formula: C₇H₈BBrO₂
MDL Number: MFCD18447597
CAS Number: 1184298-27-8

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR