Azetidine 98%

Supplier: Ambeed
Danger

A536513-1G A536513-25G A536513-250MG A536513-5G
77116-788EA 105.06 USD
77116-788 77116-782 77116-784 77116-786
Azetidine 98%
Azetidine

Caution: for Research Use Only

Formula: C₃H₇N
MDL Number: MFCD00005165
UN: 2733
ADR: 3,8,Ⅱ

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR