AZD-1981 98+%

Supplier: Ambeed
Warning

Synonyms: AZD1981, 2-(4-Acetamido-3-((4-chlorophenyl)thio)-2-methyl-1H-indol-1-yl)acetic acid

A495831-10MG A495831-1MG A495831-50MG A495831-5MG
77116-508EA 13.39 USD
77116-508 77116-510 77116-512 77116-514
AZD-1981 98+%
AZD-1981

Caution: for Research Use Only

Formula: C₁₉H₁₇ClN₂O₃S
MDL Number: MFCD26936340
CAS Number: 802904-66-1

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR