2-Phenylethanol 95%

Supplier: Ambeed
Warning

A480598-1000G A480598-100G A480598-25G A480598-500G
77116-418EA 44.28 USD
77116-418 77116-420 77116-422 77116-424
2-Phenylethanol 95%
2-Phenylethanol

Caution: for Research Use Only

Formula: C₆H₅CH₂CH₂OH
MDL Number: MFCD00002886
CAS Number: 60-12-8

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR