2,2-Dimethylbutyryl chloride 98%
Supplier: Ambeed
|
Danger
|
Synonyms:
2,2-Dimethylbutyrylchloride
A365598-25G
A365598-100G
77115-348EA
5.31
USD
77115-348
77601-616
2,2-Dimethylbutyryl chloride 98%
2,2-Dimethylbutyryl chloride
Caution: for Research Use Only
Formula:
C₆H₁₁ClO MW: 134.61 g/mol |
MDL Number:
MFCD03093071 CAS Number: 5856-77-9 UN: 2920 ADR: 8,3,Ⅱ |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...