6-(4-Chlorophenyl)pyridazin-3(2H)-one 98%
Supplier: Ambeed
|
Warning
|
A335315-25G
A335315-5G
A335315-1G
77114-966EA
43.5
USD
77114-966
77114-962
77114-964
6-(4-Chlorophenyl)pyridazin-3(2H)-one 98%
6-(4-Chlorophenyl)pyridazin-3(2H)-one
Caution: for Research Use Only
Formula:
C₁₀H₇ClN₂O |
MDL Number:
MFCD02676708 CAS Number: 2166-13-4 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...