3-Iodo-o-xylene 95%

Supplier: Ambeed
Warning

A291016-1G A291016-25G A291016-5G
77114-258EA 9.26 USD
77114-258 77114-260 77114-262
3-Iodo-o-xylene 95%
3-Iodo-o-xylene

Caution: for Research Use Only

Formula: C₈H₉I
MDL Number: MFCD00041437
CAS Number: 31599-60-7

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR