(R)-1-Methyl-3-pyrrolidinol 98%
Supplier: Ambeed
|
Warning
|
Synonyms:
(R)-3-Hydroxy-1-methylpyrrolidine, (R)-(-)-1-Methyl-3-pyrrolidinol, (R)-3-Hydroxy-1-methyl-pyrrolidine
A273989-100G
A273989-10G
A273989-1G
A273989-25G
A273989-5G
77113-938EA
593.27
USD
77113-938
77113-940
77113-942
77113-944
77113-946
(R)-1-Methyl-3-pyrrolidinol 98%
(R)-1-Methyl-3-pyrrolidinol
Caution: for Research Use Only
Formula:
C₅H₁₁NO |
MDL Number:
MFCD03788747 CAS Number: 104641-60-3 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...