3-Amino-4-fluorophenol 98%

Supplier: Ambeed
Warning

A238387-10G A238387-1G A238387-5G
77113-172EA 41.2 USD
77113-172 77113-174 77113-176
3-Amino-4-fluorophenol 98%
3-Amino-4-fluorophenol

Caution: for Research Use Only

Formula: C₆H₆FNO
MW: 127.1200 g/mol
MDL Number: MFCD09033850
CAS Number: 62257-16-3

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR