Clofarabine 97%

Supplier: Ambeed
Danger

Synonyms: 2-Chloro-2'-arabino-fluoro-2'-deoxyadenosine, (2R,3R,4S,5R)-5-(6-Amino-2-chloro-9H-purin-9-yl)-4-fluoro-2-(hydroxymethyl)tetrahydrofuran-3-ol, 2-Chloro-9-(2-deoxy-2-fluoro-beta-D-arabinofuranosyl)adenine

A195550-100MG A195550-10MG A195550-1G A195550-1MG A195550-250MG A195550-50MG
77111-956EA 45.32 USD
77111-956 77111-958 77111-960 77111-962 77111-964 77111-966
Clofarabine 97%
Clofarabine

Caution: for Research Use Only

Formula: C₁₀H₁₁ClFN₅O₃
MDL Number: MFCD00871077
CAS Number: 123318-82-1
UN: 2811
ADR: 6.1,Ⅲ

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR