L-Lysine 97% (contains < 5% H₂O)

Supplier: Ambeed
Warning

Synonyms: (S)-2,6-Diaminohexanoic acid, L(+)-Lysine, (S)-(+)-2,6-Diaminohexanoic acid, (S)-(+)-Lysine, (S)-(+)-2,6-Diaminocaproic acid, H-Lys-OH

A187445-100G A187445-10G A187445-5G A187445-25G
77111-634EA 10.61 USD
77111-634 77111-636 77111-638 77302-086
L-Lysine 97% (contains < 5% H₂O)
L-Lysine

Caution: for Research Use Only

Formula: C₆H₁₄N₂O₂
MW: 146.19 g/mol
MDL Number: MFCD00064433
CAS Number: 56-87-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR