5-Acetyl-2-chloropyridine 98%

Supplier: Ambeed
Warning

Synonyms: 3-Acetyl-6-chloropyridine

A176889-100G A176889-100MG A176889-10G A176889-1G A176889-25G A176889-5G
77111-246EA 194.66 USD
77111-246 77111-248 77111-250 77111-252 77111-254 77111-256
5-Acetyl-2-chloropyridine 98%
5-Acetyl-2-chloropyridine

Caution: for Research Use Only

Formula: C₇H₆ClNO
MW: 155.5816 g/mol
MDL Number: MFCD03407343
CAS Number: 55676-22-7

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR