4-Chloro-m-toluic acid 97%

Supplier: Ambeed
Warning

A174705-100G A174705-10G A174705-25G A174705-5G
77111-140EA 37.07 USD
77111-140 77111-142 77111-144 77111-146
4-Chloro-m-toluic acid 97%
4-Chloro-m-toluic acid
Formula: C₈H₇ClO₂
MDL Number: MFCD00045853
CAS Number: 7697-29-2

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR