4-Chlorobenzonitrile 98%

Supplier: Ambeed
Warning

Synonyms: p-Chlorobenzonitrile

A149914-1000G A149914-100G A149914-25G A149914-500G
77110-238EA 91.67 USD
77110-238 77110-240 77110-242 77110-244
4-Chlorobenzonitrile 98%
4-Chlorobenzonitrile

Caution: for Research Use Only

Formula: C₇H₄ClN
MW: 137.57 g/mol
Boiling Pt: 223
MDL Number: MFCD00001813
CAS Number: 623-03-0

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR