Isochroman 98%

Supplier: Ambeed
Warning

Synonyms: 3,4-Dihydro-1H-2-benzopyran

A148812-1000G A148812-100G A148812-10G A148812-25G A148812-500G A148812-5G
77110-192EA 328.57 USD
77110-192 77110-194 77110-196 77110-198 77110-200 77110-202
Isochroman 98%
Isochroman

Caution: for Research Use Only

Formula: C₉H₁₀O
MDL Number: MFCD00006913
CAS Number: 493-05-0

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR