Pyrogallol 97%

Supplier: Ambeed
Danger

A141606-10G A141606-25G A141606-100G
77109-822EA 9.26 USD
77109-822 77316-190 77615-130
Pyrogallol 97%
Pyrogallol

Caution: for Research Use Only

Formula: C₆H₆O₃
MDL Number: MFCD00002192
CAS Number: 87-66-1
UN: 2811
ADR: 6.1,Ⅲ

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR