Ethyl-1-phenyl-3-(trifluoromethyl)-1H-pyrazole-4-carboxylate 97%

Supplier: Ambeed
Warning

A137559-1G A137559-5G
77109-620EA 11.34 USD
77109-620 77109-622 77109-624
Ethyl-1-phenyl-3-(trifluoromethyl)-1H-pyrazole-4-carboxylate 97%
Ethyl-1-phenyl-3-(trifluoromethyl)-1H-pyrazole-4-carboxylate

Caution: for Research Use Only

Formula: C₁₃H₁₁F₃N₂O₂
MDL Number: MFCD10568308
CAS Number: 741717-63-5

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR