Methyl-4-hydroxybenzoate 98%
Supplier: Ambeed
|
Warning
|
A137460-1000G
A137460-100G
A137460-25G
A137460-500G
77109-612EA
36.04
USD
77109-612
77109-614
77109-616
77109-618
Methyl-4-hydroxybenzoate 98%
Methyl 4-hydroxybenzoate
Caution: for Research Use Only
Formula:
HOC₆H₄CO₂CH₃ |
MDL Number:
MFCD00002352 CAS Number: 99-76-3 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...