Benzocaine (ethyl-4-aminobenzoate) 98%
Supplier: Ambeed
|
Warning
|
Synonyms:
Aethoform
, Ethyl 4-aminobenzoate
A132131-50MG
A132131-100G
A132131-500G
A132131-1000G
A132131-25G
77109-306EA
62.83
USD
77109-306
77109-312
77109-310
77298-542
77109-308
Benzocaine (ethyl-4-aminobenzoate) 98%
Benzocaine (Ethyl 4-aminobenzoate)
Caution: for Research Use Only
Formula:
C₉H₁₁NO₂ MW: 165.19 g/mol |
MDL Number:
MFCD00007892 CAS Number: 94-09-7 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...