2-Methylquinoxaline 98%
Supplier: Ambeed
|
Warning
|
A115068-5G
A115068-25G
A115068-100G
77108-204EA
168.92
USD
77108-204
77108-206
77108-208
2-Methylquinoxaline 98%
2-Methylquinoxaline
Caution: for Research Use Only
Formula:
C₉H₈N₂ MW: 144.18 g/mol |
MDL Number:
MFCD00006727 CAS Number: 7251-61-8 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...