Cyanoguanidine 98%

Supplier: Ambeed
Warning

A102228-1000G A102228-100G A102228-500G
77107-238EA 20.6 USD
77107-238 77107-240 77107-242
Cyanoguanidine 98%
Cyanoguanidine

Caution: for Research Use Only

Formula: (H₂N)₂CNCN
MDL Number: MFCD00008066
CAS Number: 461-58-5

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR