PIP® G-Tek® PolyKor® FR Safety Gloves
Supplier: PIP
16-AR413/L 16-AR413/M 16-AR413/S 16-AR413/XL
76944-534DZ 171.06 USD
76944-534 76944-532 76944-530 76944-536
PIP® G-Tek® PolyKor® FR Safety Gloves
Gloves
AR seamless knit PolyKor®/Aramid blend glove with neoprene foam-coated grip on palm and fingers.
  • Cal rating of ATPV 13 cal/cm²
  • Seamless knit construction provides comfort without sacrificing dexterity
  • Microsurface neoprene coating provides a secure grip
  • EN 407 tested limited flame spread Level 4
  • ASTM F1060-08 tested contact heat resistance Level 2
PolyKor® blended shell with Aramid is lightweight and provides excellent dexterity, tactile sensitivity, and cut resistance while offering protection in heat-resistant applications, where sparks and heat are present. Neoprene foam coating provides a secure grip in dry, wet, and oily conditions, infused with thousands of tiny suction cup pockets that create a vacuum effect that dispenses fluids away on contact.