MISSION® Genomics Lentivirus Packaging Mix Powered
Supplier: Mirus Bio
MIR6640 MIR6630
76211-034EA 2219.97 USD
76211-034 76218-672
MISSION® Genomics Lentivirus Packaging Mix Powered
Transfection Reagents
Aqueous solution for enhanced lentiviral transductions in target cell types. Provided at 10 mg/ml.
  • Produce 2 to 3x higher functional titers
  • Achieve even higher titers and eliminate the need to concentrate
  • No media change required
  • Single harvest
Accelerate viral production by skipping tedious but critical multivariable plasmid optimization. Produce high titer lentivirus with the TransIT®-Lenti Transfection Reagent. Compatible with second or third generation transfer vector encoding gene of interest (GOI). Choose between small or large scale lentivirus production with our conveniently sized 5 or 34 reaction sizes for 10cm dish. Optimized formulation of two plasmids expressing the structural and regulatory (gag, pol, and rev) and viral envelope (VSV-G) proteins required for producing high titer lentivirus with broad viral tropism.