2-Tolualdehyde 96%
Supplier: Ambeed
|
Warning
|
A955800-100G
A955800-10G
A955800-25G
A955800-500G
A955800-5G
A955800-1000G
77106-408EA
13
USD
77106-408
77106-410
77106-412
77106-414
77106-416
77614-664
2-Tolualdehyde 96%
2-Tolualdehyde
Caution: for Research Use Only
Formula:
C₈H₈O |
MDL Number:
MFCD00003338 CAS Number: 529-20-4 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...