(S)-(+)-Methyl mandelate 98%

Supplier: Ambeed
Warning

Synonyms: L(+)-Mandelic acid methyl ester, L(+)-2-Hydroxy-2-phenylacetic acid methyl ester, Methyl (S)-(+)-mandelate, L(+)-Methyl mandelate, (S)-(+)-Mandelic acid methyl ester, (S)-(+)-2-Hydroxy-2-phenylacetic acid methyl ester, L(+)-Methyl glycolate, (S)-(+)-Methyl glycolate

A784791-100G A784791-1G A784791-25G A784791-5G
77105-472EA 83 USD
77105-472 77105-474 77105-476 77105-478 77105-480
(S)-(+)-Methyl mandelate 98%
(S)-(+)-Methyl mandelate

Caution: for Research Use Only

Formula: C₉H₁₀O₃
MW: 166.18 g/mol
Melting Pt: 58
MDL Number: MFCD00064246
CAS Number: 21210-43-5

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR