α,α,α-Trifluoro-o-toluonitrile 98%
Supplier: Ambeed
|
Danger
|
A630965-100G
A630965-10G
A630965-1G
A630965-25G
A630965-5G
77104-302EA
78
USD
77104-302
77104-304
77104-306
77104-308
77104-310
α,α,α-Trifluoro-o-toluonitrile 98%
α,α,α-Trifluoro-o-toluonitrile
Caution: for Research Use Only
Formula:
C₈H₄F₃N |
MDL Number:
MFCD00001791 CAS Number: 447-60-9 UN: 3276 ADR: 6.1,Ⅲ |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...