6-Methoxynicotinonitrile 98%

Supplier: Ambeed
Warning

Synonyms: 2-Methoxypyridine-5-carbonitrile

A581863-10G A581863-250MG A581863-5G A581863-1G A581863-25G
77103-794EA 43 USD
77103-794 77103-796 77103-798 77613-802 77686-479
6-Methoxynicotinonitrile 98%
6-Methoxynicotinonitrile 2-(4-Formylphenyl)isonicotinonitrile

Caution: for Research Use Only

Formula: C₇H₆N₂O
MDL Number: MFCD00084981
CAS Number: 15871-85-9

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR