α-Bromo-3-chloro-5-fluorotoluene 98%

Supplier: Ambeed
Danger

Synonyms: 1-(Bromomethyl)-3-chloro-5-fluorobenzene, 3-Chloro-5-fluorobenzyl bromide

A483145-1G A483145-5G
77102-820EA 22 USD
77102-820 77102-822
α-Bromo-3-chloro-5-fluorotoluene 98%
α-Bromo-3-chloro-5-fluorotoluene

Caution: for Research Use Only

Formula: C₇H₅BrClF
MDL Number: MFCD03788472
CAS Number: 493024-39-8
UN: 3265
ADR: 8,Ⅱ

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR