4-Iodobenzoyl chloride 98%

Supplier: Ambeed
Danger

Synonyms: p-Iodobenzoyl chloride

A427853-5G A427853-25G A427853-10G
77102-226EA 13.8 USD
77102-226 77102-222 77102-224
4-Iodobenzoyl chloride 98%
4-Iodobenzoyl chloride

Caution: for Research Use Only

Formula: C₇H₄ClIO
MW: 266.47 g/mol
MDL Number: MFCD00001058
CAS Number: 1711-02-0
UN: 3261
ADR: 8,Ⅱ

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR