Boc-S-trityl-L-homocysteine 95%

Supplier: Ambeed
Warning

Synonyms: (S)-2-((tert-Butoxycarbonyl)amino)-4-(tritylthio)butanoic acid, Boc-Homocys(Trt)-OH, Boc-HoCys(Trt)-OH

A389767-100MG A389767-250MG A389767-5G
77101-760EA 13 USD
77101-760 77101-762 77572-180
Boc-S-trityl-L-homocysteine 95%
Boc-S-trityl-L-homocysteine

Caution: for Research Use Only

Formula: C₂₈H₃₁NO₄S
MW: 477.619 g/mol
MDL Number: MFCD00672309
CAS Number: 201419-16-1

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR