Sodium ricinolate 93%

Supplier: Ambeed
Warning

A357374-100G A357374-25G A357374-500G
77106-864EA 53 USD
77106-864 77106-866 77106-868
Sodium ricinolate 93%
Sodium ricinolate

Caution: for Research Use Only

Formula: C₁₈H₃₄O₃
MDL Number: MFCD00050760
CAS Number: 5323-95-5

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR