Indoline 97%

Supplier: Ambeed
Warning

Synonyms: 2,3-Dihydroindol

A342642-1000G A342642-100G A342642-10G A342642-25G A342642-500G A342642-5G
77106-666EA 132 USD
77106-666 77106-668 77106-670 77106-672 77106-674 77106-676
Indoline 97%
Indoline

Caution: for Research Use Only

Formula: C₈H₉N
MW: 119.17 g/mol
Boiling Pt: 220
Melting Pt: -21
MDL Number: MFCD00005705
CAS Number: 496-15-1

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR