H-D-4-Pal-OH·2HCl 98%

Supplier: Ambeed
Warning

Synonyms: (R)-2-Amino-3-(4-pyridyl)propionic acid hydrochlorate, 3-(4-Pyridyl)-D-alanine hydrochlorate, 3-(4-Pyridyl)-D-alanine·2HCl, H-D-4-Pal-OH.2HCl, D-4-Pyridylalanine hydrochlorate

A339490-100MG A339490-1G A339490-5G
77106-610EA 5 USD
77106-610 77106-612 77106-614
H-D-4-Pal-OH·2HCl 98%
H-D-4-Pal-OH·2HCl

Caution: for Research Use Only

Formula: C₈H₁₀N₂O₂·₂HCl
MW: 239.2 g/mol
MDL Number: MFCD09842075
CAS Number: 174096-41-4

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR