2-Fluorothiophenol 98%
Supplier: Ambeed
|
Danger
|
Synonyms:
2-Fluorobenzenethiol, 1-Fluoro-2-mercaptobenzene, 2-Fluorobenzene-1-thiol, o-Fluorothiophenol
A318123-1G
A318123-25G
A318123-5G
77099-792EA
6
USD
77099-792
77099-794
77099-796
2-Fluorothiophenol 98%
2-Fluorothiophenol
Caution: for Research Use Only
Formula:
C₆H₅FS MW: 128.17 g/mol |
MDL Number:
MFCD00041419 CAS Number: 2557-78-0 UN: 1228 ADR: 3,6.1 Ⅲ |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...