2-Amino-6-(trifluoromethyl)pyridine 98%

Supplier: Ambeed
Danger

A228715-10G A228715-1G A228715-25G A228715-5G
77098-170EA 12 USD
77098-170 77098-172 77098-174 77098-176
2-Amino-6-(trifluoromethyl)pyridine 98%
2-Amino-6-(trifluoromethyl)pyridine

Caution: for Research Use Only

Formula: C₆H₅F₃N₂
MDL Number: MFCD03093868
CAS Number: 34486-24-3
UN: 2811
ADR: 6.1,Ⅲ

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR