Fmoc-Glu(OtBu)-OH·H₂O 98%

Supplier: Ambeed
Warning

Synonyms: (S)-2-((((9H-Fluoren-9-yl)methoxy)carbonyl)amino)-5-(tert-butoxy)-5-oxopentanoic acid hydrate

A212152-10G A212152-25G A212152-100G
77097-710EA 11.67 USD
77097-710 77097-712 77685-557
Fmoc-Glu(OtBu)-OH·H₂O 98%
Fmoc-Glu(OtBu)-OH·H₂O

Caution: for Research Use Only

Formula: C₂₄H₂₉NO₇
MW: 443.496 g/mol
MDL Number: MFCD00150485
CAS Number: 204251-24-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR