4-Bromo-2,5-difluoroaniline 98%

Supplier: Ambeed
Warning

A167178-100G A167178-10G A167178-25G A167178-500G A167178-5G
77096-358EA 98 USD
77096-358 77096-360 77096-362 77096-364 77096-366
4-Bromo-2,5-difluoroaniline 98%
4-Bromo-2,5-difluoroaniline

Caution: for Research Use Only

Formula: C₆H₄BrF₂N
MDL Number: MFCD01861125
CAS Number: 112279-60-4

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR