Sonidegib 98%

Supplier: Ambeed
Warning

Synonyms: N-(6-(cis-2,6-Dimethylmorpholino)pyridin-3-yl)-2-methyl-4'-(trifluoromethoxy)-[1,1'-biphenyl]-3-carboxamide, LDE225

A154891-1MG A154891-250MG A154891-5MG A154891-10MG A154891-1G A154891-100MG A154891-5G
77095-918EA 12 USD
77095-918 77095-920 77095-922 77611-474 77685-279 77901-006 77900-927
Sonidegib 98%
Sonidegib

Caution: for Research Use Only

Formula: C₂₆H₂₆F₃N₃O₃
MDL Number: MFCD16038928
CAS Number: 956697-53-3

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR