2-(5H-Chromeno[2,3-b]pyridin-7-yl)propanoic acid 98%

Supplier: Ambeed
Warning

Synonyms: Pranoprofen

A138748-10G A138748-1G A138748-1MG A138748-25MG A138748-5G
77095-344EA 373 USD
77095-344 77095-346 77095-348 77095-350 77095-352
2-(5H-Chromeno[2,3-b]pyridin-7-yl)propanoic acid 98%
2-(5H-Chromeno[2,3-b]pyridin-7-yl)propanoic acid

Caution: for Research Use Only

Formula: C₁₅H₁₃NO₃
MW: 255.26 g/mol
MDL Number: MFCD00866111
CAS Number: 52549-17-4

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR