18-Crown-6 (1,4,7,10,13,16-hexaoxacyclooctadecane) 98%
Supplier: Ambeed
|
Warning
|
A104233-100G
A104233-10G
A104233-25G
A104233-500G
A104233-1000G
77093-626EA
38
USD
77093-626
77093-628
77093-630
77093-632
77548-986
18-Crown-6 (1,4,7,10,13,16-hexaoxacyclooctadecane) 98%
18-Crown-6 (1,4,7,10,13,16-Hexaoxacyclooctadecane)
Caution: for Research Use Only
Formula:
C₁₂H₂₄O₆ |
MDL Number:
MFCD00005113 CAS Number: 17455-13-9 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...