HydrOasis Cinnamon Leaves
Supplier: SEGREST FARMS
HydrOasis
963-0121-9
470352-330BG 6.1 USD
470352-330
HydrOasis Cinnamon Leaves
Specimens Live Specimens Plants - Living Material
Natural decor for high impact and big benefits for your animals.
  • Recreate a naturally healthy, aquatic environment
  • Releases a natural, beneficial tint to the water
  • Creates shelter for fry, shrimp, snails, crayfish, and other invertebrates
  • Natural food sources for many bottom dwelling species and invertebrates
  • Encourages natural behavior such as hiding, grazing, and breeding
The sturdy structure and natural shape of the leaves entice your animals to show off more natural behavior as they interact in their environment by foraging, grazing and hiding. Ideal for invertebrates such as snails, shrimp, and crabs, as well as many other bottom dwelling animals.
Packaging: Simply select the desired number of leaves and place them in your aquarium. The leaves may float when first added.