ReptiFauna Jungle Leaf Litter
Supplier: SEGREST FARMS
ReptiFauna
963-0115-9
470352-320BG 10.25 USD
470352-320
ReptiFauna Jungle Leaf Litter
Specimens Live Specimens Plants - Living Material
Natural decor for high impact and big benefits for your animals.
  • Contains 5 banana leaves, 5 Catappa leaves, 5 palm leaves, and 5 guava leaves
  • Provides a soft layer for your animals to burrow and hide
  • Helps retain soil moisture and ambient humidity
  • Breaks down into nutrients for isopods and plants for a complete bioactive habitat
  • Encourages natural behavior of your animals
Leaf litter helps recreate the jungle floor and provides hiding places, to help your pet feel right at home. Assorted Leaf Litter helps support a bioactive ecosystem while your isopods and springtails benefit from the breakdown of leaf litter as a food source.
Packaging: Leaves are safe to use right out of the package. Simply select the desired amount of leaves and place them into your terrarium. Leaf litter can be scattered on top of the substrate or mixed in for the desired effect.