Magnetic Stirrers
Supplier: CRYSTAL TECHNOLOGY & INDUSTRIES
MS-01AUA MS-01DUA MS-01FUA MS-01TUA MS-01UA
76944-214EA 235.08 USD
76944-214 76944-216 76944-218 76944-220 76944-222
Magnetic Stirrers
Stirrers Magnetic Stirrers
Magnetic stirrers from Crystal Industries feature precise stirring in an ergonomical design.
  • Speeds from 0 to 2000 rpm and capacity to 1×10000 ml
  • ABS platforms provide extreme durability and chemical resistance
  • Timer with automatic stop available on MS-01DUA (0 to 99 h 59 min)
  • Five stirring power levels availabe on the MS-01UA and MS-01TUA for mixing high viscosity liquids
  • Two year replacement warranty
All models are constructed of corrosion resistant materials for easy cleaning and long service life.
Certifications: CE marked.