L(+)-Asparagine 96%

Supplier: Ambeed
Warning

A260357-25G A260357-100G A260357-500G A260357-1000G
77091-962EA 11 USD
77091-962 77113-664 77568-502 77685-741
L(+)-Asparagine 96%
L(+)-Asparagine

Caution: For research use only.

Formula: H₂NCOCH₂CH(NH₂)CO₂H
MDL Number: MFCD00064401
CAS Number: 70-47-3

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR