Guaiacol 99%

Supplier: Ambeed
Warning

Synonyms: 2-Methoxyphenol2-Hydroxyanisole1-Hydroxy-2-methoxybenzene

A242031-100G A242031-25G A242031-5G A242031-500G A242031-1000G A242031-1G
77090-420EA 8.49 USD
77090-420 77113-274 77113-272 77113-270 77113-268 77113-266
Guaiacol 99%
Guaiacol

Caution: for Research Use Only

Formula: C₇H₈O₂
MW: 124.14 g/mol
MDL Number: MFCD00002185
CAS Number: 90-05-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR